3-Chloro-4-(difluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

36026-1G 36026-25G 36026-500MG 36026-5G
76915-530EA 332.07 USD
76915-530 76915-532 76915-534 76915-536
3-Chloro-4-(difluoromethoxy)benzoic acid ≥97%
3-Chloro-4-(difluoromethoxy)benzoic acid
Formula: C₈H₅ClF₂O₃
MW: 222.573 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14652989
CAS Number: 1192829-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR