4-Amino-3-bromo-5-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

35808-1G 35808-25G 35808-5G
76915-518EA 88.06 USD
76915-518 76915-520 76915-522
4-Amino-3-bromo-5-methylbenzoic acid ≥95%
4-Amino-3-bromo-5-methylbenzoic acid
Formula: C₈H₈BrNO₂
MW: 230.059 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05975057
CAS Number: 860787-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR