4-Bromo-1-chloro-2-(difluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-1-Chloro-2-(difluoromethoxy)benzene

35787-1G 35787-25G 35787-5G
76915-504EA 111.4 USD
76915-504 76915-506 76915-508
4-Bromo-1-chloro-2-(difluoromethoxy)benzene ≥97%
4-Bromo-1-chloro-2-(difluoromethoxy)benzene
Formula: C₇H₄BrClF₂O
MW: 257.46 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09878214
CAS Number: 1000575-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR