3-Fluoro-4-(methoxymethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

35703-1G 35703-25G 35703-500MG 35703-5G
76915-488EA 315.09 USD
76915-488 76915-490 76915-492 76915-494
3-Fluoro-4-(methoxymethoxy)benzoic acid ≥97%
3-Fluoro-4-(methoxymethoxy)benzoic acid
Formula: C₉H₉FO₄
MW: 200.164 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18826051
CAS Number: 570408-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR