3-(Benzyloxy)-5-chlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

35619-1G 35619-25G 35619-5G
76915-480EA 106.09 USD
76915-480 76915-482 76915-484
3-(Benzyloxy)-5-chlorobenzaldehyde ≥97%
3-(Benzyloxy)-5-chlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16618949
CAS Number: 1228956-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR