4-Bromo-2,6-dimethyl-2H-indazole ≥97%

Supplier: AOBChem USA

Warning

35519-1G 35519-500MG 35519-5G
76915-450EA 855.09 USD
76915-450 76915-452 76915-454
4-Bromo-2,6-dimethyl-2H-indazole ≥97%
4-Bromo-2,6-dimethyl-2H-indazole
Formula: C₉H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD12028632
CAS Number: 1159511-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR