3'-Methoxy-[1,1'-biphenyl]-4-amine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3'-Methoxybiphenyl-4-ylamine, 4-(3-Methoxyphenyl)aniline

35510-25G 35510-5G
76915-440EA 389.36 USD
76915-440 76915-442
3'-Methoxy-[1,1'-biphenyl]-4-amine ≥97%
3'-Methoxy-[1,1'-biphenyl]-4-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO
MW: 199.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03424684
CAS Number: 207287-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR