3-(4-Nitrophenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

35506-1G 35506-25G 35506-500MG 35506-5G
76915-424EA 215.37 USD
76915-424 76915-426 76915-428 76915-430
3-(4-Nitrophenyl)pyridine ≥97%
3-(4-Nitrophenyl)pyridine
Formula: C₁₁H₈N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00276489
CAS Number: 4282-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR