4,4'-Dichlorobiphenyl (PCB No. 15) ≥97%

Supplier: AOBChem USA

Warning

Synonyms: PCB No. 15, 4,4'-PCB, 4,4'-Dichloro-1,1'-biphenyl

35242-25G 35242-5G
76915-376EA 590.93 USD
76915-376 76915-378
4,4'-Dichlorobiphenyl (PCB No. 15) ≥97%
4,4'-Dichlorobiphenyl (PCB No. 15)

Precaución: for Research Use Only

Formula: C₁₂H₈Cl₂
MW: 223.1 g/mol
Storage Temperature: Ambient
CAS Number: 2050-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR