3-(2-Pyridyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-(Pyridin-2-yl)benzaldehyde

35120-1G 35120-25G 35120-5G
76915-320EA 64.72 USD
76915-320 76915-322 76915-324
3-(2-Pyridyl)benzaldehyde ≥95%
3-(2-Pyridyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₉NO
Storage Temperature: Refrigerator
MDL Number: MFCD02684103
CAS Number: 85553-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR