3-(Thien-3-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

34866-1G 34866-25G 34866-5G
76915-252EA 112.46 USD
76915-252 76915-254 76915-256
3-(Thien-3-yl)benzaldehyde ≥97%
3-(Thien-3-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₈OS
Storage Temperature: Refrigerator
MDL Number: MFCD04039150
CAS Number: 129746-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR