3,4-Difluoro-2-hydroxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

34766-1G 34766-25G 34766-500MG 34766-5G
76915-190EA 374.5 USD
76915-190 76915-192 76915-194 76915-196
3,4-Difluoro-2-hydroxyphenylboronic acid pinacol ester ≥97%
3,4-Difluoro-2-hydroxyphenylboronic acid pinacol ester
Formula: C₁₂H₁₅BF₂O₃
MW: 256.054 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414855
CAS Number: 1631756-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR