4-Bromo-2-(thiomethyl)thiazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-(methylthio)thiazole

34738-1G 34738-25G 34738-5G
76915-180EA 76.39 USD
76915-180 76915-182 76915-184
4-Bromo-2-(thiomethyl)thiazole ≥97%
4-Bromo-2-(thiomethyl)thiazole
Formula: C₄H₄BrNS₂
MW: 210.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07787376
CAS Number: 204513-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR