3-Bromo-2-(4-methoxyphenyl)pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

34679-1G 34679-500MG 34679-5G
76915-152EA 635.48 USD
76915-152 76915-154 76915-156
3-Bromo-2-(4-methoxyphenyl)pyridine ≥97% research grade
3-Bromo-2-(4-methoxyphenyl)pyridine
Formula: C₁₂H₁₀BrNO
Storage Temperature: Refrigerator
MDL Number: MFCD23143943
CAS Number: 1171902-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR