3-(p-Tolyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(4-Methylphenyl)pyridine

34599-1G 34599-25G 34599-500MG 34599-5G
76915-136EA 171.87 USD
76915-136 76915-138 76915-140 76915-142
3-(p-Tolyl)pyridine ≥97%
3-(p-Tolyl)pyridine
Formula: C₁₂H₁₁N
MW: 169.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038775
CAS Number: 4423-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR