3,5-Difluoro-4-formylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

34539-1G 34539-25G 34539-5G
76915-126EA 76.39 USD
76915-126 76915-128 76915-130
3,5-Difluoro-4-formylphenylboronic acid pinacol ester ≥97%
3,5-Difluoro-4-formylphenylboronic acid pinacol ester
Formula: C₁₃H₁₅BF₂O₃
MW: 268.064 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17215216
CAS Number: 2088247-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR