3-Chloro-4-fluoro-5-iodobenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

34241-1G 34241-25G 34241-500MG 34241-5G
76914-974EA 337.37 USD
76914-974 76914-976 76914-978 76914-980
3-Chloro-4-fluoro-5-iodobenzotrifluoride ≥97%
3-Chloro-4-fluoro-5-iodobenzotrifluoride
Formula: C₇H₂ClF₄I
Storage Temperature: Refrigerator
MDL Number: MFCD31619474
CAS Number: 126538-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR