4-(Cyclopropylmethoxy)-3-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

34201-1G 34201-25G 34201-500MG 34201-5G
76914-958EA 245.07 USD
76914-958 76914-960 76914-962 76914-964
4-(Cyclopropylmethoxy)-3-fluorobenzaldehyde ≥97%
4-(Cyclopropylmethoxy)-3-fluorobenzaldehyde
Formula: C₁₁H₁₁FO₂
MW: 194.202 g/mol
Storage Temperature: Refrigerator
CAS Number: 883525-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR