4-(6-Methoxypyridin-2-yl)benzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

34195-1G 34195-500MG 34195-5G
76914-948EA 485.9 USD
76914-948 76914-950 76914-952
4-(6-Methoxypyridin-2-yl)benzaldehyde ≥97% research grade
4-(6-Methoxypyridin-2-yl)benzaldehyde
Formula: C₁₃H₁₁NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD05150363
CAS Number: 618092-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR