3,5-Dichloro-2-methyl-benzoic acid ≥97%

Supplier: AOBChem USA

Warning

34098-1G 34098-500MG 34098-5G
76914-934EA 448.77 USD
76914-934 76914-936 76914-938
3,5-Dichloro-2-methyl-benzoic acid ≥97%
3,5-Dichloro-2-methyl-benzoic acid
Formula: C₈H₆Cl₂O₂
MW: 205.038 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22495123
CAS Number: 101567-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR