4-Bromo-2,3-Dichlorophenol ≥97%

Supplier: AOBChem USA

Warning

34078-1G 34078-25G 34078-5G
76914-922EA 84.88 USD
76914-922 76914-924 76914-926
4-Bromo-2,3-Dichlorophenol ≥97%
4-Bromo-2,3-Dichlorophenol
Formula: C₆H₃BrCl₂O
MW: 241.90 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00152292
CAS Number: 1940-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR