3-Bromo-2,4-dichlorotoluene ≥97%

Supplier: AOBChem USA

Warning

34011-25G 34011-5G
76914-912EA 495.45 USD
76914-912 76914-914
3-Bromo-2,4-dichlorotoluene ≥97%
3-Bromo-2,4-dichlorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂
Storage Temperature: Ambient
MDL Number: MFCD00079712
CAS Number: 206559-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR