3-Chloro-2-fluoro-4-methylpyridine ≥97% research grade

Supplier: AOBChem USA

Warning

33889-1G 33889-500MG 33889-5G
76914-898EA 585.62 USD
76914-898 76914-900 76914-902
3-Chloro-2-fluoro-4-methylpyridine ≥97% research grade
3-Chloro-2-fluoro-4-methylpyridine
Formula: C₆H₅ClFN
Storage Temperature: Ambient
MDL Number: MFCD13185446
CAS Number: 1214377-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR