3-Isopropylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

33701-25G 33701-5G
76914-876EA 559.1 USD
76914-876 76914-878
3-Isopropylbenzaldehyde ≥97%
3-Isopropylbenzaldehyde
Formula: C₁₀H₁₂O
MW: 148.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062435
CAS Number: 34246-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR