3-Cyano-2,4-dichlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

33699-1G 33699-500MG 33699-5G
76914-862EA 485.9 USD
76914-862 76914-864 76914-866
3-Cyano-2,4-dichlorobenzoic acid ≥97%
3-Cyano-2,4-dichlorobenzoic acid
Formula: C₈H₃Cl₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD28758617
CAS Number: 1807162-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR