3-Bromo-5-(trifluoromethoxy)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

33686-25G 33686-5G
76914-850EA 630.18 USD
76914-850 76914-852
3-Bromo-5-(trifluoromethoxy)benzyl alcohol ≥97%
3-Bromo-5-(trifluoromethoxy)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆BrF₃O₂
MW: 271.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09999512
CAS Number: 1026201-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR