3-Bromo-2,5-dichlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

33673-1G 33673-500MG 33673-5G
76914-836EA 500.75 USD
76914-836 76914-838 76914-840
3-Bromo-2,5-dichlorobenzaldehyde ≥97%
3-Bromo-2,5-dichlorobenzaldehyde
Formula: C₇H₃BrCl₂O
MW: 253.908 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28166251
CAS Number: 1823608-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR