3-Bromo-4-phenyl-2-chloropyridine ≥95% research grade
Supplier: AOBChem USA
|
Warning
|
33619-1G
33619-25G
33619-500MG
33619-5G
76914-804EA
350.1
USD
76914-804
76914-806
76914-808
76914-810
3-Bromo-4-phenyl-2-chloropyridine ≥95% research grade
3-Bromo-4-phenyl-2-chloropyridine
Formula:
C₁₁H₇BrClN Storage Temperature: Refrigerator |
MDL Number:
MFCD28123459 CAS Number: 1622838-93-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...