3-(Benzyloxy)-5-bromobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

33599-1G 33599-25G 33599-500MG 33599-5G
76914-782EA 124.13 USD
76914-782 76914-784 76914-786 76914-788
3-(Benzyloxy)-5-bromobenzoic acid ≥97%
3-(Benzyloxy)-5-bromobenzoic acid
Formula: C₁₄H₁₁BrO₃
MW: 307.139 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17015777
CAS Number: 1242336-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR