3-Chloro-5-fluoro-2-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

33536-1G 33536-25G 33536-500MG 33536-5G
76914-750EA 439.22 USD
76914-750 76914-752 76914-754 76914-756
3-Chloro-5-fluoro-2-methoxybenzoic acid ≥95%
3-Chloro-5-fluoro-2-methoxybenzoic acid
Formula: C₈H₆ClFO₃
MW: 204.583 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22415153
CAS Number: 1554882-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR