3-Chloro-5-fluoro-2-methoxyphenol ≥97% research grade

Supplier: AOBChem USA

Warning

33534-1G 33534-25G 33534-500MG 33534-5G
76914-734EA 296 USD
76914-734 76914-736 76914-738 76914-740
3-Chloro-5-fluoro-2-methoxyphenol ≥97% research grade
3-Chloro-5-fluoro-2-methoxyphenol
Formula: C₇H₆ClFO₂
MW: 176.573 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22415068
CAS Number: 1783971-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR