3-Bromo-2-fluoro-5-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

33498-1G 33498-25G 33498-500MG 33498-5G
76914-692EA 180.36 USD
76914-692 76914-694 76914-696 76914-698
3-Bromo-2-fluoro-5-(trifluoromethyl)benzyl alcohol ≥97%
3-Bromo-2-fluoro-5-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₅BrF₄O
MW: 273.022 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30834069
CAS Number: 2092564-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR