3,5-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3,5-Dichloropyridine-4-boronic acid pinacol ester

33384-1G 33384-25G 33384-5G
76914-670EA 62.6 USD
76914-670 76914-672 76914-674
3,5-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97% research grade
3,5-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Formula: C₁₁H₁₄BCl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD12546037
CAS Number: 1257641-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR