3-Chloro-2-methoxy-5-nitropyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
33253-1G
33253-25G
33253-5G
76914-656EA
84.88
USD
76914-656
76914-658
76914-660
3-Chloro-2-methoxy-5-nitropyridine ≥97%
3-Chloro-2-methoxy-5-nitropyridine
Precaución: for Research Use Only
Formula:
C₆H₅ClN₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD18254096 CAS Number: 22353-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...