3,4-Difluoro-5-ethoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

33183-1G 33183-25G 33183-500MG 33183-5G
76914-640EA 201.58 USD
76914-640 76914-642 76914-644 76914-646
3,4-Difluoro-5-ethoxyphenylboronic acid ≥97%
3,4-Difluoro-5-ethoxyphenylboronic acid
Formula: C₈H₉BF₂O₃
MW: 201.963 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26404349
CAS Number: 1162261-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR