3,4-Difluoro-5-methoxybenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
33179-1G
33179-25G
33179-5G
76914-626EA
62.6
USD
76914-626
76914-628
76914-630
3,4-Difluoro-5-methoxybenzaldehyde ≥97%
3,4-Difluoro-5-methoxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₆F₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD04972450 CAS Number: 881190-46-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...