3,4-Difluoro-5-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(3,4-Difluoro-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,4-Difluoro-5-methoxyphenylboronic acid, pinacol ester

33177-1G 33177-25G 33177-500MG 33177-5G
76914-610EA 207.94 USD
76914-610 76914-612 76914-614 76914-616
3,4-Difluoro-5-methoxyphenylboronic acid pinacol ester ≥97%
3,4-Difluoro-5-methoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BF₂O₃
MW: 270.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15143617
CAS Number: 754226-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR