(3,4-Dichloro-2-methoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

33151-1G 33151-25G 33151-5G
76914-598EA 94.43 USD
76914-598 76914-600 76914-602
(3,4-Dichloro-2-methoxyphenyl)boronic acid ≥97%
(3,4-Dichloro-2-methoxyphenyl)boronic acid
Formula: C₇H₇BCl₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD18434427
CAS Number: 2096329-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR