3-Bromo-5-nitrobenzamide ≥97%

Supplier: AOBChem USA

Warning

33104-25G 33104-5G
76914-580EA 300.24 USD
76914-580 76914-582
3-Bromo-5-nitrobenzamide ≥97%
3-Bromo-5-nitrobenzamide

Precaución: for Research Use Only

Formula: C₇H₅BrN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD01529620
CAS Number: 54321-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR