3-Formyl-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

32961-1G 32961-25G 32961-500MG 32961-5G
76914-550EA 210.06 USD
76914-550 76914-552 76914-554 76914-556
3-Formyl-2-methoxybenzoic acid ≥97%
3-Formyl-2-methoxybenzoic acid
Formula: C₉H₈O₄
MW: 180.157 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24387285
CAS Number: 1289170-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR