3-Chloro-2-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32955-1G 32955-25G 32955-500MG 32955-5G
76914-528EA 303.42 USD
76914-528 76914-530 76914-532 76914-534
3-Chloro-2-isopropoxybenzaldehyde ≥97%
3-Chloro-2-isopropoxybenzaldehyde
Formula: C₁₀H₁₁ClO₂
MW: 198.646 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24686241
CAS Number: 894851-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR