3-Chloro-2-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

32953-1G 32953-25G 32953-500MG 32953-5G
76914-512EA 346.92 USD
76914-512 76914-514 76914-516 76914-518
3-Chloro-2-methoxybenzyl alcohol ≥97%
3-Chloro-2-methoxybenzyl alcohol
Formula: C₈H₉ClO₂
MW: 172.609 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12755469
CAS Number: 869088-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR