2-(3-(Methoxymethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(Methoxymethyl)phenylboronic acid, pinacol ester

32926-1G 32926-25G 32926-5G
76914-488EA 84.88 USD
76914-488 76914-490 76914-492
2-(3-(Methoxymethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(3-(Methoxymethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₄H₂₁BO₃
MW: 248.126 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06795646
CAS Number: 675605-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR