3,5-Difluoro-4-methylanisole ≥95%

Supplier: AOBChem USA

Warning

32676-25G 32676-5G
76914-440EA 453.01 USD
76914-440 76914-442
3,5-Difluoro-4-methylanisole ≥95%
3,5-Difluoro-4-methylanisole
Formula: C₈H₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD18395267
CAS Number: 1261478-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR