3,5-Bis(trifluoromethyl)-2-hydroxybenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
32665-1G
32665-500MG
32665-5G
76914-426EA
530.45
USD
76914-426
76914-428
76914-430
3,5-Bis(trifluoromethyl)-2-hydroxybenzoic acid ≥97%
3,5-Bis(trifluoromethyl)-2-hydroxybenzoic acid
Formula:
C₉H₄F₆O₃ MW: 274.117 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28730981 CAS Number: 197015-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...