4-(Benzyloxy)-3-chloro-5-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

32651-1G 32651-25G 32651-500MG 32651-5G
76914-410EA 267.35 USD
76914-410 76914-412 76914-414 76914-416
4-(Benzyloxy)-3-chloro-5-fluorophenol ≥97%
4-(Benzyloxy)-3-chloro-5-fluorophenol
Formula: C₁₃H₁₀ClFO₂
MW: 252.669 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29044728
CAS Number: 1881289-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR