3-Chloro-2,4-dimethylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

32630-1G 32630-25G 32630-500MG 32630-5G
76914-352EA 337.37 USD
76914-352 76914-354 76914-356 76914-358
3-Chloro-2,4-dimethylphenylboronic acid ≥97%
3-Chloro-2,4-dimethylphenylboronic acid
Formula: C₈H₁₀BClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD30529832
CAS Number: 2121515-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR