3-Bromoquinolin-2(1H)-one ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-1H-quinolin-2-one

32618-1G 32618-25G 32618-500MG 32618-5G
76914-338EA 131.56 USD
76914-338 76914-340 76914-342 76914-344
3-Bromoquinolin-2(1H)-one ≥97%
3-Bromoquinolin-2(1H)-one
Formula: C₉H₆BrNO
MW: 224.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09029798
CAS Number: 939-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR