3-Chloro-2-ethoxy-4-formylpyridine ≥97%

Supplier: AOBChem USA

Warning

32611-1G 32611-25G 32611-500MG 32611-5G
76914-326EA 337.37 USD
76914-326 76914-328 76914-330 76914-332
3-Chloro-2-ethoxy-4-formylpyridine ≥97%
3-Chloro-2-ethoxy-4-formylpyridine
Formula: C₈H₈ClNO₂
MW: 185.608 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30529827
CAS Number: 2056110-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR