3-Chloro-4-iodo-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

32599-1G 32599-25G 32599-5G
76914-298EA 117.76 USD
76914-298 76914-300 76914-302
3-Chloro-4-iodo-2-methoxypyridine ≥97%
3-Chloro-4-iodo-2-methoxypyridine
Formula: C₆H₅ClINO
Storage Temperature: Refrigerator
MDL Number: MFCD16610527
CAS Number: 1227603-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR